siRNA Details
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012392.1 | NP_001012392.1 | 13, 14 | 1365 | ||
NM_153418.2 | NP_700467.2 | 13, 14 | 1328 | |||
GenBank mRNA | BC060982.1 | - | 13, 14 | 1365 | 30300112 | |
BC100436.1 | - | 13, 14 | 1360 | 30298932 | ||
U46068.3 | - | 13, 14 | 1329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
