siRNA Details
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004721.1 | NP_001004721.1 | 4 | 318 | ||
GenBank mRNA | AK140955.1 | - | 4 | 314 | ||
AK143517.1 | - | 4 | 318 | |||
AK162913.1 | - | 293 | ||||
AK171988.1 | - | 4 | 297 | |||
AK211799.1 | - | 2 | 98 | |||
BB839941.1 | - | 4 | 318 | |||
BC120680.1 | - | 4 | 307 | 40129603 | ||
BC120682.1 | - | 4 | 307 | 40129605 | ||
CK334159.1 | - | 4 | 271 |

More Information
Other Species Targeted

Gene Ontology Categories:
