siRNA Details
Gene Symbol: Dlgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042487.1 | NP_001035952.1 | 7 | 2234 | ||
NM_001042488.2 | NP_001035953.1 | 7 | 1533 | |||
NM_001277186.1 | NP_001264115.1 | 13 | 3469 | |||
NM_001277187.1 | NP_001264116.1 | 12 | 3427 | |||
NM_146128.6 | NP_666240.4 | 13 | 3478 | |||
GenBank mRNA | AK122408.1 | - | 11 | 6136 | ||
AK150554.1 | - | 7 | 1454 | |||
AK180044.1 | - | 1 | 127 | |||
AK186004.1 | - | 1 | 126 | |||
BC024558.1 | - | 7 | 1638 | |||
BC058948.1 | - | 7 | 1557 | |||
BC141110.1 | - | 11 | 3106 | 9055722 | ||
BC145394.1 | - | 11 | 3106 | 9053952 | ||
BC145395.1 | - | 10 | 3064 | 9053953 |
