siRNA Details
Gene Symbol: D630003M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131021.2 | NP_001124493.1 | 3 | 1030 | ||
NM_177657.5 | NP_808325.2 | 3 | 1030 | |||
GenBank mRNA | AK052605.1 | - | 3 | 1029 | ||
AK168367.1 | - | 3 | 1032 | |||
BC137959.1 | - | 3 | 941 | 8860978 | ||
BC137960.1 | - | 3 | 941 | 8860979 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
