siRNA Details
Gene Symbol: Ppp1r16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159662.1 | NP_001153134.1 | 10 | 1287 | ||
NM_153089.4 | NP_694729.1 | 10 | 1265 | |||
GenBank mRNA | AF423761.1 | - | 10 | 1284 | ||
AK037627.1 | - | 1582 | ||||
AK042123.1 | - | 10 | 1262 | |||
AK044453.1 | - | 10 | 1303 |

More Information
Other Species Targeted

Gene Ontology Categories:
