siRNA Details
Gene Symbol: Gdap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144891.2 | NP_659140.2 | 2 | 383 | ||
GenBank mRNA | AK140488.1 | - | 2 | 374 | ||
AK141371.1 | - | 2 | 375 | |||
AK164991.1 | - | 2 | 434 | |||
BB649122.1 | - | 2 | 399 | |||
BC019941.1 | - | 2 | 397 | 3969548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
