siRNA Details
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252587.2 | NP_001239516.1 | 3, 4 | 318 | ||
NM_001291158.1 | NP_001278087.1 | 3, 4 | 2240 | |||
NM_027230.5 | NP_081506.3 | 3, 4 | 318 | |||
GenBank mRNA | AB093284.2 | - | 3, 4 | 2232 | ||
AK080667.1 | - | 3, 4 | 824 | |||
BC048186.1 | - | 3, 4 | 125 | 6827541 | ||
BC063267.1 | - | 2, 3 | 99 | |||
CN670238.1 | - | 3, 4 | 133 |
