siRNA Details
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153065.3 | NP_694705.2 | 11 | 1233 | ||
GenBank mRNA | AK134274.1 | - | 11 | 1234 | ||
AK138442.1 | - | 11 | 1257 | |||
AK146469.1 | - | 11 | 1261 | |||
BC011321.1 | - | 10 | 1110 | 4007058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
