siRNA Details
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 10 | 1132 | ||
GenBank mRNA | AK048537.1 | - | 10 | 1052 | ||
AK145292.1 | - | 10 | 1164 | |||
AK161695.1 | - | 10 | 1071 | |||
AK166059.1 | - | 10 | 1111 | |||
AK170306.1 | - | 10 | 1116 | |||
AK170669.1 | - | 10 | 1133 | |||
AK178638.1 | - | 4 | 311 | |||
AK178810.1 | - | 4 | 362 | |||
AK182769.1 | - | 4 | 320 | |||
AK220257.1 | - | 4 | 451 | |||
BC026804.1 | - | 10 | 1139 | 4159217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
