siRNA Details
Gene Symbol: Tmem189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145538.2 | NP_663513.1 | 5 | 558 | ||
GenBank mRNA | AK088196.1 | - | 5 | 571 | ||
AK151326.1 | - | 544 | ||||
AK170042.1 | - | 568 | ||||
AK179462.1 | - | 2 | 156 | |||
AK212647.1 | - | 1 | 25 | |||
AK213753.1 | - | 3 | 244 | |||
BC002270.1 | - | 5 | 558 | 3496088 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
