siRNA Details
Gene Symbol: Eif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005509.2 | NP_001005509.1 | 5 | 596 | ||
GenBank mRNA | AK050971.1 | - | 5 | 313 | ||
AK078519.1 | - | 5 | 375 | |||
AK145275.1 | - | 5 | 552 | |||
AK161904.1 | - | 5 | 377 | |||
BC030447.1 | - | 4 | 265 | 5400637 | ||
BC082557.1 | - | 5 | 401 | 30606936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
