siRNA Details
Gene Symbol: Syt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018804.3 | NP_061274.2 | 4 | 1428 | ||
GenBank mRNA | AB026808.1 | - | 1153 | |||
AK122200.1 | - | 3 | 922 | |||
AK133414.1 | - | 1856 | ||||
AK144169.1 | - | 4 | 1346 | |||
AK158745.1 | - | 4 | 1324 | |||
AK164368.1 | - | 4 | 1378 | |||
BC025207.1 | - | 3 | 646 | 5030173 | ||
BC054526.1 | - | 4 | 1369 | 5698412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
