siRNA Details
Gene Symbol: Pias3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165949.1 | NP_001159421.1 | 9 | 1224 | ||
NM_018812.2 | NP_061282.2 | 10 | 1119 | |||
NM_146135.2 | NP_666247.1 | 9 | 1261 | |||
GenBank mRNA | AF034080.3 | - | 9 | 996 | ||
AK040619.1 | - | 9 | 1252 | |||
AK077551.1 | - | 9 | 1210 | |||
AK085503.1 | - | 9 | 1196 | |||
AK134404.1 | - | 4 | 399 | |||
BC023128.1 | - | 9 | 1167 | 3585764 | ||
BC050988.1 | - | 10 | 1078 | |||
BC051252.1 | - | 9 | 1134 | 6489803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
