siRNA Details
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146136.1 | NP_666248.1 | 8, 9 | 1514 | ||
GenBank mRNA | AK045797.1 | - | 8, 9 | 1500 | ||
AK049873.1 | - | 8, 9 | 1577 | |||
AK085492.1 | - | 8, 9 | 1511 | |||
AK136837.1 | - | 8, 9 | 1513 | |||
BC026596.1 | - | 8, 9 | 1514 | 4975010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
