siRNA Details
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146136.1 | NP_666248.1 | 3 | 341 | ||
GenBank mRNA | AK045797.1 | - | 3 | 327 | ||
AK049873.1 | - | 3 | 404 | |||
AK052779.1 | - | 3 | 340 | |||
AK085492.1 | - | 3 | 338 | |||
AK136837.1 | - | 3 | 340 | |||
BC025441.1 | - | 3 | 329 | 5043709 | ||
BC026596.1 | - | 3 | 341 | 4975010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
