siRNA Details
Gene Symbol: Strip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153563.2 | NP_705791.2 | 11 | 1367 | ||
GenBank mRNA | AK032065.1 | - | 11 | 1349 | ||
AK122553.1 | - | 11 | 1331 | |||
AK148011.1 | - | 11 | 1352 | |||
AK169644.1 | - | 11 | 1299 | |||
AK185112.1 | - | 1 | 17 | |||
AK187609.1 | - | 1 | 17 | |||
AK197998.1 | - | 1 | 17 | |||
AK205847.1 | - | 1 | 17 | |||
BC023952.1 | - | 11 | 1343 | 5320305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
