siRNA Details
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080818.2 | NP_001074287.1 | 2 | 644 | ||
NM_001173553.1 | NP_001167024.1 | 2 | 644 | |||
GenBank mRNA | AK032515.1 | - | 1 | 1333 | ||
AK043956.1 | - | 2 | 628 | |||
AK048250.1 | - | 2 | 549 | |||
AK052560.1 | - | 2 | 635 | |||
AK136546.1 | - | 2 | 103 | |||
AK142294.1 | - | 2 | 644 | |||
BC072644.1 | - | 2 | 598 | 30356962 |

More Information
Other Species Targeted

Gene Ontology Categories:
