siRNA Details
Gene Symbol: Lppr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177664.5 | NP_808332.3 | 6 | 1520 | ||
GenBank mRNA | AF541279.1 | - | 6 | 1521 | ||
AK046782.1 | - | 6 | 1521 | |||
AK079635.1 | - | 6 | 854 | |||
AK129149.1 | - | 6 | 1116 | |||
AY266266.1 | - | 6 | 1521 | |||
AY266267.1 | - | 6 | 1521 | |||
BC082323.1 | - | 5 | 2498 | |||
BC137701.1 | - | 6 | 1115 | 8860720 | ||
BC137702.1 | - | 6 | 1116 | 8860721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
