siRNA Details
Gene Symbol: Nol6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139236.3 | NP_631982.2 | 17 | 2306 | ||
GenBank mRNA | AF361077.1 | - | 17 | 2255 | ||
AF361078.1 | - | 17 | 2255 | |||
AK089138.1 | - | 17 | 2228 | |||
AK153714.1 | - | 978 | ||||
AK155818.1 | - | 17 | 2245 | |||
BC019981.1 | - | 3 | 261 | |||
BC021856.1 | - | 3 | 311 | |||
BC059820.1 | - | 17 | 2224 | 6827596 | ||
BC060189.1 | - | 17 | 2234 |

More Information
Other Species Targeted

Gene Ontology Categories:
