siRNA Details
Gene Symbol: N28178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172690.2 | NP_766278.1 | 6 | 784 | ||
GenBank mRNA | AK038528.1 | - | 5 | 638 | ||
AK046283.1 | - | 6 | 775 | |||
AK122428.1 | - | 4 | 707 | |||
BC057092.1 | - | 6 | 784 | 5703966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
