siRNA Details
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173788.3 | NP_776149.1 | 16 | 2669 | ||
GenBank mRNA | AK158195.1 | - | 11 | 1220 | ||
BC023420.1 | - | 3 | 310 | |||
BC037458.1 | - | 11 | 1125 | |||
BC042470.1 | - | 16 | 2669 | 5137261 | ||
BC049896.1 | - | 6 | 764 |
