siRNA Details
Gene Symbol: Zfp189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289901.1 | NP_001276830.1 | 4 | 568 | ||
NM_145547.3 | NP_663522.2 | 3 | 739 | |||
GenBank mRNA | AK049475.1 | - | 3 | 648 | ||
AK051265.1 | - | 3 | 698 | |||
BC021326.1 | - | 3 | 701 | 5065942 | ||
BC087833.1 | - | 3 | 605 | 5708354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
