siRNA Details
Gene Symbol: Ikbkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026079.3 | NP_080355.2 | 17 | 2069 | ||
GenBank mRNA | AF140786.1 | - | 17 | 1997 | ||
AF367244.1 | - | 17 | 2022 | |||
AF387811.1 | - | 17 | 2036 | |||
AK031367.1 | - | 17 | 2014 | |||
AK035944.1 | - | 17 | 2071 | |||
AK147153.1 | - | 17 | 2072 | |||
AK165168.1 | - | 14 | 1546 | |||
BC052387.1 | - | 17 | 1981 | 5705589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
