siRNA Details
Gene Symbol: E130308A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015681.2 | NP_001015681.1 | 2 | 538 | ||
NM_153158.5 | NP_694798.1 | 2 | 538 | |||
GenBank mRNA | AK047478.1 | - | 2 | 539 | ||
AK047601.1 | - | 2 | 507 | |||
AK081604.1 | - | 2 | 422 | |||
AK087509.1 | - | 2 | 535 | |||
BC017649.1 | - | 2 | 305 | 4924105 | ||
BC037070.1 | - | 2 | 356 | 5004205 |

More Information
Other Species Targeted

Gene Ontology Categories:
