siRNA Details
Gene Symbol: Ifne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177348.2 | NP_796322.1 | 1 | 112 | ||
GenBank mRNA | AY190044.1 | - | 1 | 112 | ||
BC104376.2 | - | 1 | 111 | 40050053 | ||
BC104377.2 | - | 1 | 111 | 40050054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
