siRNA Details
Gene Symbol: Cyp2j13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145548.4 | NP_663523.2 | 5 | 807 | ||
GenBank mRNA | AK143903.1 | - | 5 | 808 | ||
AK143981.1 | - | 5 | 804 | |||
AK162358.1 | - | 811 | ||||
AK165405.1 | - | 5 | 807 | |||
AK165469.1 | - | 5 | 785 | |||
AK165473.1 | - | 5 | 785 | |||
BC024451.1 | - | 4 | 427 |
