siRNA Details
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301414.1 | NP_001288343.1 | 3 | 777 | ||
NM_146144.4 | NP_666256.2 | 3 | 758 | |||
GenBank mRNA | AK080882.1 | - | 3 | 759 | ||
BC018179.1 | - | 3 | 638 | 3585191 | ||
BC020007.1 | - | 3 | 610 | 3980469 |

More Information
Other Species Targeted

Gene Ontology Categories:
