siRNA Details
Gene Symbol: Fam151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146149.1 | NP_666261.1 | 4, 5 | 627 | ||
GenBank mRNA | AY168619.1 | - | 4, 5 | 627 | ||
BC024426.1 | - | 4, 5 | 627 | 4971419 | ||
BC024430.1 | - | 4, 5 | 627 | 4972368 | ||
BC026682.1 | - | 4, 5 | 636 | 5065146 |
