siRNA Details
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146151.4 | NP_666263.3 | 10 | 1354 | ||
GenBank mRNA | AK031814.1 | - | 10 | 1347 | ||
AK047755.1 | - | 10 | 1355 | |||
AK134575.1 | - | 6 | 460 | |||
AK211719.1 | - | 1 | 22 | |||
BC019149.1 | - | 9 | 1087 | 5053345 | ||
BC029766.1 | - | 10 | 1332 | 4947564 |
