siRNA Details
Gene Symbol: AU022252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012400.2 | NP_001012400.1 | 3 | 301 | ||
GenBank mRNA | AK158349.1 | - | 3 | 259 | ||
AK167445.1 | - | 3 | 263 | |||
BC035522.1 | - | 3 | 284 | |||
BC089614.1 | - | 3 | 283 | 6748514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
