siRNA Details
Gene Symbol: AU022252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012400.2 | NP_001012400.1 | 4 | 397 | ||
GenBank mRNA | AK167445.1 | - | 4 | 359 | ||
BC035522.1 | - | 4 | 380 | |||
BC089614.1 | - | 4 | 379 | 6748514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
