siRNA Details
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256432.1 | NP_001243361.1 | 3 | 425 | ||
NM_177671.5 | NP_808339.2 | 3 | 425 | |||
GenBank mRNA | AK039700.1 | - | 3 | 388 | ||
AK143168.1 | - | 1310 | ||||
BC116991.1 | - | 3 | 355 | 40126310 | ||
BC119084.1 | - | 3 | 355 | 8733833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
