siRNA Details
Gene Symbol: 5730409E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013755.3 | NP_001013777.2 | 2 | 257 | ||
NM_001145950.1 | NP_001139422.1 | 2 | 262 | |||
GenBank mRNA | AK077438.1 | - | 2 | 247 | ||
BY123366.1 | - | 2 | 299 |
