siRNA Details
Gene Symbol: Fam167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182783.2 | NP_877584.2 | 2 | 794 | ||
GenBank mRNA | AK134621.1 | - | 2 | 624 | ||
AK143928.1 | - | 2 | 764 | |||
BC030183.1 | - | 2 | 761 | |||
BX517339.1 | - | 2 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
