siRNA Details
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039677.2 | NP_001034766.1 | 8 | 1298 | ||
GenBank mRNA | AK031425.1 | - | 8 | 1095 | ||
BC013478.1 | - | 7 | 1318 | |||
BC113751.1 | - | 8 | 1298 | 40057211 | ||
BC147704.1 | - | 8 | 1129 | 9087463 | ||
BC147707.1 | - | 8 | 1129 | 9087466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
