siRNA Details
Gene Symbol: Ncmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168498.1 | NP_001161970.1 | 5 | 1051 | ||
NM_001168500.1 | NP_001161972.1 | 5 | 1253 | |||
NM_001243305.1 | NP_001230234.1 | 4 | 864 | |||
NM_001243306.1 | NP_001230235.1 | 6 | 1421 | |||
NM_145555.2 | NP_663530.1 | 5 | 1250 | |||
NM_181047.1 | NP_851390.1 | 5 | 1048 | |||
GenBank mRNA | AK014757.1 | - | 5 | 1027 | ||
AK039485.1 | - | 4 | 893 | |||
AK076488.1 | - | 5 | 1002 | |||
AK167453.1 | - | 5 | 1251 | |||
BC005730.1 | - | 6 | 1416 | 3595770 | ||
BC037722.1 | - | 5 | 1227 | |||
BC039748.1 | - | 6 | 1427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
