siRNA Details
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198610.2 | NP_941012.1 | 8, 9 | 1728 | ||
GenBank mRNA | AK137439.1 | - | 8, 9 | 1727 | ||
BC055811.1 | - | 8, 9 | 1701 | 6414125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
