siRNA Details
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008725.2 | NP_032751.1 | 3 | 699 | ||
GenBank mRNA | AK147180.1 | - | 3 | 703 | ||
BC089615.1 | - | 3 | 699 | 6755676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
