siRNA Details
Gene Symbol: Tmem201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284270.1 | NP_001271199.1 | 2 | 287 | ||
NM_001284273.1 | NP_001271202.1 | 2 | 274 | |||
NM_177672.4 | NP_808340.2 | 2 | 287 | |||
GenBank mRNA | AK039221.1 | - | 2 | 217 | ||
AK078532.1 | - | 2 | 213 | |||
AK079157.1 | - | 2 | 194 | |||
AK086543.1 | - | 2 | 264 | |||
AK153631.1 | - | 2 | 264 | |||
AK159041.1 | - | 2 | 275 | |||
BC072645.1 | - | 2 | 253 | 30357476 | ||
BY233050.1 | - | 2 | 265 | |||
CD353644.1 | - | 2 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
