siRNA Details
Gene Symbol: Tmem201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284270.1 | NP_001271199.1 | 2 | 303 | ||
NM_001284273.1 | NP_001271202.1 | 2 | 290 | |||
NM_177672.4 | NP_808340.2 | 2 | 303 | |||
GenBank mRNA | AK039221.1 | - | 2 | 233 | ||
AK078532.1 | - | 2 | 229 | |||
AK079157.1 | - | 2 | 210 | |||
AK086543.1 | - | 2 | 280 | |||
AK153631.1 | - | 2 | 280 | |||
AK159041.1 | - | 2 | 291 | |||
BC072645.1 | - | 2 | 269 | 30357476 | ||
BY233050.1 | - | 2 | 281 | |||
CD353644.1 | - | 2 | 171 |
