siRNA Details
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089983.1 | - | 6 | 606 | ||
AK156793.1 | - | 5 | 1818 | |||
AK171781.1 | - | 6 | 595 | |||
AK172542.1 | - | 6 | 595 | |||
BC085257.1 | - | 6 | 588 | 5414963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
