siRNA Details
Gene Symbol: 9430015G10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145557.3 | NP_663532.2 | 4 | 265 | ||
NM_177205.1 | NP_796179.1 | 4 | 415 | |||
GenBank mRNA | AK029276.1 | - | 4 | 279 | ||
AK034620.1 | - | 4 | 225 | |||
BC004010.1 | - | 4 | 285 | 3493145 | ||
BQ830972.1 | - | 250 | ||||
BY727027.1 | - | 4 | 394 | |||
CO803636.1 | - | 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
