siRNA Details
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 6 | 871 | ||
GenBank mRNA | AK034358.1 | - | 6 | 903 | ||
AK036245.1 | - | 6 | 947 | |||
AK045969.1 | - | 6 | 905 | |||
AK048738.1 | - | 6 | 857 | |||
AK079644.1 | - | 5 | 802 | |||
AK082474.1 | - | 5 | 816 | |||
AK133610.1 | - | 6 | 872 | |||
AK134808.1 | - | 6 | 932 | |||
AK161902.1 | - | 6 | 932 | |||
BC132290.1 | - | 6 | 847 | 40130567 | ||
BC138012.1 | - | 6 | 847 | 8861031 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109345.
Gene Ontology Categories:
