siRNA Details
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 4 | 610 | ||
GenBank mRNA | AK034358.1 | - | 4 | 642 | ||
AK036245.1 | - | 4 | 686 | |||
AK043026.1 | - | 4 | 674 | |||
AK045969.1 | - | 4 | 644 | |||
AK048738.1 | - | 4 | 630 | |||
AK079644.1 | - | 3 | 541 | |||
AK081041.1 | - | 4 | 655 | |||
AK082474.1 | - | 3 | 555 | |||
AK133610.1 | - | 4 | 611 | |||
AK134808.1 | - | 4 | 671 | |||
AK161902.1 | - | 4 | 671 | |||
AK165601.1 | - | 4 | 664 | |||
BC132290.1 | - | 4 | 586 | 40130567 | ||
BC138012.1 | - | 4 | 586 | 8861031 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
