siRNA Details
Gene Symbol: Nupl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153092.4 | NP_694732.3 | 7 | 842 | ||
GenBank mRNA | AB067574.1 | - | 6 | 690 | ||
AK077066.1 | - | 7 | 810 | |||
AK078478.1 | - | 7 | 843 | |||
BC033270.1 | - | 7 | 784 | 3981211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
