siRNA Details
Gene Symbol: 4931409K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177676.6 | NP_808344.3 | 18 | 2317 | ||
GenBank mRNA | AK029839.1 | - | 17 | 2749 | ||
AV271477.2 | - | 2 | 263 | |||
BC137999.1 | - | 14 | 2038 | 8861018 | ||
BC145792.1 | - | 14 | 2040 | 40131095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
