siRNA Details
Gene Symbol: Tnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139064.2 | NP_620703.1 | 3 | 715 | ||
GenBank mRNA | AJ304865.1 | - | 3 | 715 | ||
AK160165.1 | - | 3 | 673 | |||
BC052083.1 | - | 4 | 761 | 6404610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
