siRNA Details
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194344.2 | NP_919325.2 | 10 | 1541 | ||
GenBank mRNA | AK028482.1 | - | 6 | 1637 | ||
AK131143.1 | - | 4 | 406 | |||
AK150064.1 | - | 10 | 1319 | |||
AK155330.1 | - | 10 | 1303 | |||
AK159089.1 | - | 10 | 1320 | |||
AK167283.1 | - | 10 | 1242 | |||
AK170383.1 | - | 10 | 1374 | |||
AK170396.1 | - | 10 | 1321 | |||
AK171209.1 | - | 10 | 1298 | |||
AK171242.1 | - | 10 | 1298 | |||
BC024909.1 | - | 8 | 991 | |||
BC056461.1 | - | 8 | 885 |

More Information
Other Species Targeted

Gene Ontology Categories:
