siRNA Details
Gene Symbol: Cytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081106.1 | NP_001074575.1 | 4 | 656 | ||
GenBank mRNA | AK076770.1 | - | 3 | 548 | ||
BC063103.1 | - | 4 | 616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
