siRNA Details
Gene Symbol: Atp10d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153389.3 | NP_700438.3 | 2 | 476 | ||
GenBank mRNA | AJ441079.1 | - | 2 | 476 | ||
AK036646.1 | - | 1 | 141 | |||
AK087333.1 | - | 2 | 492 | |||
BC099527.1 | - | 2 | 466 | |||
BC141328.1 | - | 2 | 327 | 9055940 |

More Information
Other Species Targeted

Gene Ontology Categories:
